Tisk…

Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 4C12]

Dodavatel: OriGene
ORIGCF505827EA 21120 CZK
ORIGCF505827
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 4C12]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3R5
Clonality: Monoclonal
Clone: 4C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR