Tisk…

Anti-PFKP Mouse Monoclonal Antibody [clone: 3A12]

Dodavatel: OriGene
ORIGCF503980EA 21120 CZK
ORIGCF503980
Anti-PFKP Mouse Monoclonal Antibody [clone: 3A12]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PFKP
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR