Tisk…

Anti-RYBP Mouse Monoclonal Antibody [clone: 4E10]

Dodavatel: OriGene
ORIGCF504761EA 21120 CZK
ORIGCF504761
Anti-RYBP Mouse Monoclonal Antibody [clone: 4E10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RYBP
Clonality: Monoclonal
Clone: 4E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR