Tisk…
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1F9]
Dodavatel: OriGene
ORIGCF504798EA 21120 CZK
ORIGCF504798
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1F9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human