Tisk…

Anti-PHF7 Mouse Monoclonal Antibody [clone: 1C2]

Dodavatel: OriGene
ORIGCF505156EA 21120 CZK
ORIGCF505156
Anti-PHF7 Mouse Monoclonal Antibody [clone: 1C2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PHF7
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR