Tisk…

Anti-SDSL Mouse Monoclonal Antibody [clone: 1G4]

Dodavatel: OriGene
ORIGCF505180EA 21120 CZK
ORIGCF505180
Anti-SDSL Mouse Monoclonal Antibody [clone: 1G4]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: SDSL
Clonality: Monoclonal
Clone: 1G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR