Tisk…

Anti-UBE2S Mouse Monoclonal Antibody [clone: 4H3]

Dodavatel: OriGene
ORIGCF505187EA 21120 CZK
ORIGCF505187
Anti-UBE2S Mouse Monoclonal Antibody [clone: 4H3]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: UBE2S
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR