Tisk…

Anti-LZIC Mouse Monoclonal Antibody [clone: 1D5]

Dodavatel: OriGene
ORIGCF505895EA 21710 CZK
ORIGCF505895
Anti-LZIC Mouse Monoclonal Antibody [clone: 1D5]
Protilátky
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150

Type: Primary
Antigen: LZIC
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR