Tisk…

Anti-RNF39 Mouse Monoclonal Antibody [clone: 5E10]

Dodavatel: OriGene
ORIGCF505912EA 21120 CZK
ORIGCF505912
Anti-RNF39 Mouse Monoclonal Antibody [clone: 5E10]
Protilátky
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RNF39
Clonality: Monoclonal
Clone: 5E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR