Tisk…

Anti-TPSG1 Mouse Monoclonal Antibody [clone: 2C3]

Dodavatel: OriGene
ORIGCF504028EA 21120 CZK
ORIGCF504028
Anti-TPSG1 Mouse Monoclonal Antibody [clone: 2C3]
Protilátky
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TPSG1
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR