Tisk…

Anti-DPP9 Mouse Monoclonal Antibody [clone: 6A12]

Dodavatel: OriGene
ORIGCF504039EA 21120 CZK
ORIGCF504039
Anti-DPP9 Mouse Monoclonal Antibody [clone: 6A12]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP9
Clonality: Monoclonal
Clone: 6A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR