Tisk…

Anti-SYNJ2BP Mouse Monoclonal Antibody [clone: 3A9]

Dodavatel: OriGene
ORIGCF504051EA 21120 CZK
ORIGCF504051
Anti-SYNJ2BP Mouse Monoclonal Antibody [clone: 3A9]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SYNJ2BP
Clonality: Monoclonal
Clone: 3A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR