Tisk…

Anti-MLF1 Mouse Monoclonal Antibody [clone: 3E7]

Dodavatel: OriGene
ORIGCF504827EA 21120 CZK
ORIGCF504827
Anti-MLF1 Mouse Monoclonal Antibody [clone: 3E7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MLF1
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR