Tisk…

Anti-MLF2 Mouse Monoclonal Antibody [clone: 1C4]

Dodavatel: OriGene
ORIGCF504845EA 21710 CZK
ORIGCF504845
Anti-MLF2 Mouse Monoclonal Antibody [clone: 1C4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MLF2
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR