Tisk…

Anti-OTUB1 Mouse Monoclonal Antibody [clone: 5F6]

Dodavatel: OriGene
ORIGCF505198EA 21120 CZK
ORIGCF505198
Anti-OTUB1 Mouse Monoclonal Antibody [clone: 5F6]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: OTUB1
Clonality: Monoclonal
Clone: 5F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR