Tisk…

Anti-PARVA Mouse Monoclonal Antibody [clone: 2B11]

Dodavatel: OriGene
ORIGCF506000EA 21120 CZK
ORIGCF506000
Anti-PARVA Mouse Monoclonal Antibody [clone: 2B11]
Protilátky
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PARVA
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR