Tisk…

Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 6C6]

Dodavatel: OriGene
ORIGCF506031EA 21120 CZK
ORIGCF506031
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 6C6]
Protilátky
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR