Tisk…

Anti-AK4 Mouse Monoclonal Antibody [clone: 5B10]

Dodavatel: OriGene
ORIGCF502899EA 21120 CZK
ORIGCF502899
Anti-AK4 Mouse Monoclonal Antibody [clone: 5B10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: AK4
Clonality: Monoclonal
Clone: 5B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR