Tisk…

Anti-PDE1B Mouse Monoclonal Antibody [clone: 6B9]

Dodavatel: OriGene
ORIGCF503325EA 21120 CZK
ORIGCF503325
Anti-PDE1B Mouse Monoclonal Antibody [clone: 6B9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE1B
Clonality: Monoclonal
Clone: 6B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR