Tisk…

Anti-SETD7 Mouse Monoclonal Antibody [clone: 5B11]

Dodavatel: OriGene
ORIGCF503354EA 21120 CZK
ORIGCF503354
Anti-SETD7 Mouse Monoclonal Antibody [clone: 5B11]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: SETD7
Clonality: Monoclonal
Clone: 5B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR