Tisk…

Anti-IFI35 Mouse Monoclonal Antibody [clone: 1E3]

Dodavatel: OriGene
ORIGCF503362EA 21710 CZK
ORIGCF503362
Anti-IFI35 Mouse Monoclonal Antibody [clone: 1E3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: IFI35
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR