Tisk…
Anti-GOLM1 Mouse Monoclonal Antibody [clone: 2F3]
Dodavatel: OriGene
ORIGCF504115EA 21120 CZK
ORIGCF504115
Anti-GOLM1 Mouse Monoclonal Antibody [clone: 2F3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: GOLM1
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human