Tisk…

Anti-RNF113B Mouse Monoclonal Antibody [clone: 2H5]

Dodavatel: OriGene
ORIGCF504118EA 21120 CZK
ORIGCF504118
Anti-RNF113B Mouse Monoclonal Antibody [clone: 2H5]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RNF113B
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR