Tisk…

Anti-TPSG1 Mouse Monoclonal Antibody [clone: 2A10]

Dodavatel: OriGene
ORIGCF504128EA 21120 CZK
ORIGCF504128
Anti-TPSG1 Mouse Monoclonal Antibody [clone: 2A10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TPSG1
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR