Tisk…

Anti-MGLL Mouse Monoclonal Antibody [clone: 1H1]

Dodavatel: OriGene
ORIGCF502932EA 21120 CZK
ORIGCF502932
Anti-MGLL Mouse Monoclonal Antibody [clone: 1H1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MGLL
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR