Tisk…

Anti-RAB30 Mouse Monoclonal Antibody [clone: 3E7]

Dodavatel: OriGene
ORIGCF505344EA 21120 CZK
ORIGCF505344
Anti-RAB30 Mouse Monoclonal Antibody [clone: 3E7]
Protilátky
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RAB30
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR