Tisk…

Anti-ZSCAN18 Mouse Monoclonal Antibody [clone: 2A9]

Dodavatel: OriGene
ORIGCF505348EA 21710 CZK
ORIGCF505348
Anti-ZSCAN18 Mouse Monoclonal Antibody [clone: 2A9]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: ZSCAN18
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR