Tisk…

Anti-ATG3 Mouse Monoclonal Antibody [clone: 2B9]

Dodavatel: OriGene
ORIGCF503384EA 21120 CZK
ORIGCF503384
Anti-ATG3 Mouse Monoclonal Antibody [clone: 2B9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATG3
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR