Tisk…

Anti-TPSG1 Mouse Monoclonal Antibody [clone: 1G1]

Dodavatel: OriGene
ORIGCF504161EA 21120 CZK
ORIGCF504161
Anti-TPSG1 Mouse Monoclonal Antibody [clone: 1G1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: TPSG1
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR