Tisk…

Anti-NMT2 Mouse Monoclonal Antibody [clone: 1D7]

Dodavatel: OriGene
ORIGCF504200EA 21710 CZK
ORIGCF504200
Anti-NMT2 Mouse Monoclonal Antibody [clone: 1D7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NMT2
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR