Tisk…

Anti-PDF Mouse Monoclonal Antibody [clone: 3C12]

Dodavatel: OriGene
ORIGCF502989EA 21710 CZK
ORIGCF502989
Anti-PDF Mouse Monoclonal Antibody [clone: 3C12]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDF
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR