Tisk…

Anti-TSC22D3 Mouse Monoclonal Antibody [clone: 3F3]

Dodavatel: OriGene
ORIGCF505357EA 21120 CZK
ORIGCF505357
Anti-TSC22D3 Mouse Monoclonal Antibody [clone: 3F3]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: TSC22D3
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR