Tisk…

Anti-NKIRAS1 Mouse Monoclonal Antibody [clone: 8A3]

Dodavatel: OriGene
ORIGCF505395EA 21120 CZK
ORIGCF505395
Anti-NKIRAS1 Mouse Monoclonal Antibody [clone: 8A3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NKIRAS1
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR