Tisk…

Anti-CCM2 Mouse Monoclonal Antibody [clone: 1D1]

Dodavatel: OriGene
ORIGCF503419EA 21120 CZK
ORIGCF503419
Anti-CCM2 Mouse Monoclonal Antibody [clone: 1D1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CCM2
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR