Tisk…

Anti-PGAM2 Mouse Monoclonal Antibody [clone: 4E9]

Dodavatel: OriGene
ORIGCF503425EA 21120 CZK
ORIGCF503425
Anti-PGAM2 Mouse Monoclonal Antibody [clone: 4E9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PGAM2
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR