Tisk…

Anti-AKT1 Mouse Monoclonal Antibody [clone: 2B9]

Dodavatel: OriGene
ORIGCF504236EA 21120 CZK
ORIGCF504236
Anti-AKT1 Mouse Monoclonal Antibody [clone: 2B9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR