Tisk…

Anti-AP2M1 Mouse Monoclonal Antibody [clone: 1B11]

Dodavatel: OriGene
ORIGCF503020EA 21120 CZK
ORIGCF503020
Anti-AP2M1 Mouse Monoclonal Antibody [clone: 1B11]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AP2M1
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR