Tisk…

Anti-STAT4 Mouse Monoclonal Antibody [clone: 4F9]

Dodavatel: OriGene
ORIGCF503022EA 21710 CZK
ORIGCF503022
Anti-STAT4 Mouse Monoclonal Antibody [clone: 4F9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT4
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR