Tisk…

Anti-NDEL1 Mouse Monoclonal Antibody [clone: 1G10]

Dodavatel: OriGene
ORIGCF503058EA 21120 CZK
ORIGCF503058
Anti-NDEL1 Mouse Monoclonal Antibody [clone: 1G10]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NDEL1
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR