Tisk…

Anti-GADD45G Mouse Monoclonal Antibody [clone: 2A12]

Dodavatel: OriGene
ORIGCF505441EA 21120 CZK
ORIGCF505441
Anti-GADD45G Mouse Monoclonal Antibody [clone: 2A12]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: GADD45G
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR