Tisk…

Anti-PCDH7 Mouse Monoclonal Antibody [clone: 3C8]

Dodavatel: OriGene
ORIGCF505451EA 21120 CZK
ORIGCF505451
Anti-PCDH7 Mouse Monoclonal Antibody [clone: 3C8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PCDH7
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR