Tisk…

Anti-CAPN2 Mouse Monoclonal Antibody [clone: 3G1]

Dodavatel: OriGene
ORIGCF504280EA 21120 CZK
ORIGCF504280
Anti-CAPN2 Mouse Monoclonal Antibody [clone: 3G1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CAPN2
Clonality: Monoclonal
Clone: 3G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR