Tisk…

Anti-LENG1 Mouse Monoclonal Antibody [clone: 7E10]

Dodavatel: OriGene
ORIGCF503089EA 21710 CZK
ORIGCF503089
Anti-LENG1 Mouse Monoclonal Antibody [clone: 7E10]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LENG1
Clonality: Monoclonal
Clone: 7E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR