Tisk…

Anti-TOMM34 Mouse Monoclonal Antibody [clone: 2A9]

Dodavatel: OriGene
ORIGCF503090EA 21120 CZK
ORIGCF503090
Anti-TOMM34 Mouse Monoclonal Antibody [clone: 2A9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: TOMM34
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR