Tisk…

Anti-FNDC4 Mouse Monoclonal Antibody [clone: 3E10]

Dodavatel: OriGene
ORIGCF505459EA 21710 CZK
ORIGCF505459
Anti-FNDC4 Mouse Monoclonal Antibody [clone: 3E10]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: FNDC4
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR