Tisk…

Anti-ERBB2 Mouse Monoclonal Antibody [clone: 11A7]

Dodavatel: OriGene
ORIGCF503524EA 21120 CZK
ORIGCF503524
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 11A7]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 11A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR