Tisk…

Anti-WWTR1 Mouse Monoclonal Antibody [clone: 1H9]

Dodavatel: OriGene
ORIGCF503538EA 21710 CZK
ORIGCF503538
Anti-WWTR1 Mouse Monoclonal Antibody [clone: 1H9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: WWTR1
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR