Tisk…

Anti-EPN2 Mouse Monoclonal Antibody [clone: 1E4]

Dodavatel: OriGene
ORIGCF504302EA 21710 CZK
ORIGCF504302
Anti-EPN2 Mouse Monoclonal Antibody [clone: 1E4]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: EPN2
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR