Tisk…

Anti-DPP9 Mouse Monoclonal Antibody [clone: 4C10]

Dodavatel: OriGene
ORIGCF504304EA 21120 CZK
ORIGCF504304
Anti-DPP9 Mouse Monoclonal Antibody [clone: 4C10]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: DPP9
Clonality: Monoclonal
Clone: 4C10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR