Tisk…

Anti-PSMD3 Mouse Monoclonal Antibody [clone: 2A2]

Dodavatel: OriGene
ORIGCF504337EA 21120 CZK
ORIGCF504337
Anti-PSMD3 Mouse Monoclonal Antibody [clone: 2A2]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PSMD3
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR